Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KRBA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565620
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565620 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565620 Supplier Invitrogen™ Supplier No. PA565620
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DLQGLSRGTAR RARPLPPDAP PAEPPGLHCS SSQQLLSSTP SCHAAPPAHP LLAHTGGHQS PLPPLVPAAL PLQGASPPAA SADADVPTSG VAPDGIPERP KEPSSLLGGV QRALQEELWG GEHRDPRW Highest antigen sequence identity to the following orthologs - mouse 50%, rat 48%.

KRBA1 gene ontology annotations related to this gene include regulation of transcription, DNA-templated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KRBA1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KRBA1
Gene Accession No. A5PL33
Gene Alias KIAA1862; KRAB-A domain containing 1; KRBA1; Protein KRBA1
Gene Symbols KRBA1
Host Species Rabbit
Immunogen Recombinant Human KRBA1. Recombinant protein control fragment (Product #RP-104903).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84626
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.