Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KRBA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554358
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554358 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554358 Supplier Invitrogen™ Supplier No. PA554358
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: INCLKEILVP GPRHPETSPS FLPPLPSLGT SRLTRADLGP GSPPWAVKTE AVSGDCPLQG LLHCLKELPE AQDRHPSPSG VGNRRLQENP GAWKRGSGGP GYLLTPPPHP DLGAG Highest antigen sequence identity to the following orthologs: Mouse - 61%, Rat - 59%.

KRBA1 gene ontology annotations related to this gene include regulation of transcription, DNA-templated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KRBA1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KRBA1
Gene Accession No. A5PL33
Gene Alias KIAA1862; KRAB-A domain containing 1; KRBA1; Protein KRBA1
Gene Symbols KRBA1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KRBA1. Recombinant protein control fragment (Product #RP-92721).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84626
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.