Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KRBOX4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584688
Description
Immunogen sequence: VDEQIDHYKE SQDKLPWQAA FIGKETLKDE SGQ.
This encodes a zinc finger protein with an N-terminal KRAB (Kruppel-associated) domain found in transcriptional repressors. This gene is located in a region of the X chromosome thought to be involved in nonsyndromic X-linked mental retardation. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Aug 2011].
Specifications
KRBOX4 | |
Polyclonal | |
Unconjugated | |
KRBOX4 | |
KRAB box domain containing 4; KRAB box domain-containing protein 4; KRAB domain-containing protein 4; KRBOX4; putative zinc finger transcription factor; zinc finger family member 673; zinc finger protein 673; ZNF673 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55634 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5JUW0 | |
KRBOX4 | |
Recombinant protein corresponding to Human KRBOX4. Recombinant protein control fragment (Product #RP-105351). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction