Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KRBOX4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598479
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598479 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598479 Supplier Invitrogen™ Supplier No. PA598479
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KRBOX4 Polyclonal Antibody for Western Blot, ICC/IF, ELISA

This encodes a zinc finger protein with an N-terminal KRAB (Kruppel-associated) domain found in transcriptional repressors. This gene is located in a region of the X chromosome thought to be involved in nonsyndromic X-linked mental retardation. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Aug 2011].
TRUSTED_SUSTAINABILITY

Specifications

Antigen KRBOX4
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene KRBOX4
Gene Accession No. Q5JUW0
Gene Alias KRAB box domain containing 4; KRAB box domain-containing protein 4; KRAB domain-containing protein 4; KRBOX4; putative zinc finger transcription factor; zinc finger family member 673; zinc finger protein 673; ZNF673
Gene Symbols KRBOX4
Host Species Rabbit
Immunogen Recombinant Human KRAB domain-containing protein 4 protein (1-98AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55634
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.