Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ KRCA 0008
SDP

Catalog No. 509850R Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-985-0R 50 mg
50-981-0R 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 50-985-0R Supplier Tocris Bioscience™ Supplier No. 5098/50

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent Ack1 and ALK dual inhibitor; orally bioavailable

KRCA 0008 is a potent Ack1 and anaplastic lymphoma kinase (ALK) dual inhibitor (IC50 values are 4 and 12 nM respectively). Inhibits lung cancer H3122 cell proliferation (IC50 = 80 nM). Attenuates H3122 cell xenograft tumor growth in mice. Orally bioavailable.

Specifications

Purity >99%
Quantity 50 mg
Product Type KRCA 0008
Molecular Weight (g/mol) 609.12
Inhibitors KRCA 0008

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.