Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KRCC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544658
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544658 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544658 Supplier Invitrogen™ Supplier No. PA544658
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: KMKGDYLETC GYKGEVNSRP TYRMFDQRLP SETIQTYPRS CNIPQTVENR Sequence homology: Horse: 77%; Human: 100%.

KRCC1 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KRCC1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene KRCC1
Gene Accession No. Q9NPI7
Gene Alias AA792894; BM-003; BM-044; CHBP2; cryptogenic hepatitis-binding protein 2; KRCC1; lysine rich coiled-coil 1; lysine-rich coiled-coil 1; lysine-rich coiled-coil protein 1; RGD1306495
Gene Symbols KRCC1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human KRCC1 (aa 35-84).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51315
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.