Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRCC1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989166
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9166 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9166 Supplier Bioss Supplier No. BS9560R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KRCC1 is a protein coding gene.

Specifications

Antigen KRCC1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene KRCC1
Gene Accession No. Q9NPI7
Gene Alias BM-003; BM-044; CHBP2; Cryptogenic hepatitis-binding protein 2; KRCC1; lysine rich coiled-coil 1; lysine-rich coiled-coil 1; Lysine-rich coiled-coil protein 1
Gene Symbols KRCC1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human KRCC1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51315
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.