Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kremen-1 Antibody (RM0120-6K45), DyLight 350, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP122498UV
Description
Kremen-1 Monoclonal specifically detects Kremen-1 in Mouse samples. It is validated for Western Blot.Specifications
Kremen-1 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
KREMEN1 | |
Mouse recombinant protein of the extracellular domain of Kremen-1 | |
0.1 mL | |
Primary | |
This was produced from a hybridoma (mouse myeloma) fused with spleen cells from a rat immunized with mouse recombinant protein of the extracellular domain of Kremen-1. | |
Store at 4C in the dark. | |
IgG2 |
Western Blot | |
RM0120-6K45 | |
Western Blot | |
EC 3.4.21, FLJ31863, KREMEM1, KREMEN, kremen protein 1, kringle containing transmembrane protein, kringle containing transmembrane protein 1, Kringle domain-containing transmembrane protein 1, kringle-coding gene marking the eye and the nose, Kringle-containing protein marking the eye and the nose, KRM1Dickkopf receptor | |
Rat | |
Protein A or G purified | |
Signal Transduction | |
83999 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction