Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KREMEN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547176
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547176 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547176 Supplier Invitrogen™ Supplier No. PA547176
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs, approximately 15% cross-reactivity with recombinant human Kremen-2 and recombinant mouse Kremen-2 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

KREMEN1 is a high-affinity dickkopf homolog 1 (DKK1) transmembrane receptor that functionally cooperates with DKK1 to block wingless (WNT)/beta-catenin signaling. This protein is a component of a membrane complex that modulates canonical WNT signaling through lipoprotein receptor-related protein 6 (LRP6). It contains extracellular kringle, WSC, and CUB domains.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KREMEN1
Applications Flow Cytometry, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Kremen1
Gene Accession No. Q99N43
Gene Alias AV002070; dickkopf receptor; KREMEN; kremen protein 1; KREMEN1; kringle containing transmembrane protein 1; kringle domain-containing transmembrane protein 1; kringle-coding gene marking the eye and the nose; kringle-containing protein marking the eye and the nose; KRM1
Gene Symbols Kremen1
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Kremen-1 Ala20-Gly395.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84035
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.