Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KREMEN2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547997
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547997 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547997 Supplier Thermo Scientific Supplier No. PA547997
Add to Cart
Add to Cart

KREMEN2 Polyclonal Antibody

KREMEN2 Polyclonal Antibody for Western Blot, Flow

TRUSTED_SUSTAINABILITY

Specifications

Antigen KREMEN2
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene KREMEN2
Gene Accession No. Q8NCW0
Gene Alias 2900054E04Rik; Dickkopf receptor 2; kremen protein 2; KREMEN2; kringle containing transmembrane protein 2; kringle domain-containing transmembrane protein 2; Kringle-containing protein marking the eye and the nose; Krm2
Gene Symbols KREMEN2
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Kremen-2 Gln19-Ala364.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79412
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.