Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KREMEN2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 381700
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-170-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 38-170-0 Supplier Invitrogen™ Supplier No. 381700
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KREMEN2 Polyclonal Antibody for Western Blot

Kremen (Kringle containing protein marking the eye and the nose) proteins are type I transmembrane proteins that contain extracellular kringle, WSC and CUB domains and an intracellular region without any conserved motifs. Kremens bind a subset of the secreted Dickkopf proteins (Dkk 1, 2, and 4) with high affinity to modulate the canonical Wnt signaling pathway that is transduced by the ternary receptor complex composed of Wnt, Frizzled, and the LDL receptor related protein 5/6 (LRP5/6) co-receptor. Recent experiments have shown that KREMEN2 is a regulator of bone remodeling and raise the possibility that antagonizing KREMEN2 might prove beneficial in patients with bone loss disorders.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KREMEN2
Applications Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene KREMEN2
Gene Accession No. Q8K1S7
Gene Alias 2900054E04Rik; Dickkopf receptor 2; kremen protein 2; KREMEN2; kringle containing transmembrane protein 2; kringle domain-containing transmembrane protein 2; Kringle-containing protein marking the eye and the nose; Krm2
Gene Symbols KREMEN2
Host Species Rabbit
Immunogen Synthetic peptide derived from the internal region of human, mouse and rat Kremen2 proteins.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 73016
Target Species Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.