Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRG2/DDX11 Polyclonal Antibody, Bioss
SDP

Catalog No. 502279446
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-9446 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-9446 Supplier Bioss Supplier No. BS22217R
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

KRG2/DDX11 Polyclonal antibody specifically detects KRG2/DDX11 in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen KRG2/DDX11
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300
Gene DDX11
Gene Accession No. Q6AXC6, Q96FC9
Gene Alias 4732462I11Rik; ATP-dependent DNA helicase DDX11; CHL1; CHL1-like helicase homolog; CHL1-related helicase gene-1; CHL1-related protein 1; CHLR1; DDX11; DEAD/H (Asp-Glu-Ala-Asp/His) box helicase 11; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 11 (CHL1-like helicase homolog, S. cerevisiae); DEAD/H box protein 11; DEAD/H-box helicase 11; DEAD/H-box protein 11; essa15a; hCHLR1; Keratinocyte growth factor-regulated gene 2 protein; KRG2; KRG-2; probable ATP-dependent DNA helicase DDX11; probable ATP-dependent RNA helicase DDX11; WABS
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human KRG2/DDX11
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1663, 316767, 320209
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.