Learn More
KRI1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304640A

Description
The recommended shelf life for this product is 1 year from date of receipt.
This gene overlaps with the gene for cysteineendopeptidase AUT-like 4 in a head-to-tail orientation.Specifications
KRI1 | |
Polyclonal | |
Unconjugated | |
KRI1 | |
protein KRI1 homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
65095 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q8N9T8 | |
KRI1 | |
Between 200 and 250 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.