Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KRIT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562020
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562020 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562020 Supplier Invitrogen™ Supplier No. PA562020
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SLQLKPYHKP LQHVRDWPEI LAELTNLDPQ RETPQLFLRR DVRLPLEVEK QIEDPLAILI LFDEARYNLL KGFYTAPDAK LITLASLLLQ IVYGNYESKK HKQGFLNEEN LKSIV Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 97%.

This gene encodes a protein containing four ankyrin repeats, a band 4.1/ezrin/radixin/moesin (FERM) domain, and multiple NPXY sequences. The encoded protein is localized in the nucleus and cytoplasm. It binds to integrin cytoplasmic domain-associated protein-1 alpha (ICAP1alpha), and plays a critical role in beta1-integrin-mediated cell proliferation. It associates with junction proteins and RAS-related protein 1A (Rap1A), which requires the encoded protein for maintaining the integrity of endothelial junctions. It is also a microtubule-associated protein and may play a role in microtubule targeting. Mutations in this gene result in cerebral cavernous malformations. Multiple alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KRIT1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KRIT1
Gene Accession No. O00522
Gene Alias 2010007K12Rik; A630036P20Rik; AA432855; AI450393; AI643869; ankyrin repeat-containing protein Krit1; BB155247; BB235701; CAM; Ccm1; Cerebral cavernous malformations 1 protein; cerebral cavernous malformations 1 protein homolog; Krev interaction trapped 1; krev interaction trapped protein 1; KRIT1; KRIT1, ankyrin repeat containing; RGD1305929
Gene Symbols KRIT1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KRIT1. Recombinant protein control fragment (Product #RP-92586).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 889
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.