Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KRIT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP305054100UL
Description
KRIT1 Polyclonal specifically detects KRIT1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
KRIT1 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
889 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunofluorescence | |
Unconjugated | |
CAMcerebral cavernous malformations 1, CCM1ankyrin repeat-containing protein Krit1, Cerebral cavernous malformations 1 protein, Krev interaction trapped 1, krev interaction trapped protein 1, KRIT1, ankyrin repeat containing | |
Recombinant fusion protein containing a sequence corresponding to amino acids 467-736 of human KRIT1 (NP_919438.1). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction