Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Kromasil C18 HPLC Column, 100 Å, 5 μm Particle Size

Catalog No. 111003530 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
125 mm
150 mm
250 mm
Diameter (Metric):
4 mm
4.6 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
11-100-3530 125 mm 4 mm
11-101-6068 150 mm 4.6 mm
11-102-0737 250 mm 4.6 mm
11-100-3414 250 mm 4 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 11-100-3530 Supplier MilliporeSigma™ Supelco™ Supplier No. K08670658
Add to Cart
Add to Cart

Kromasil C18 HPLC Column, 100 Å, 5 μm Particle Size for small molecules and peptides.

Kromasil Classic 100 Å family of products is used to separate and purify molecules of up to about 10,000 Da.

  • Kromasil™ C18 HPLC column involves a silica-based packing.
  • The reduced levels of Na, K, Ca, Fe, Al, and Mg make the column useful for polar compounds, which tend to interact with the silica surface.
  • Reverse phase (RP) HPLC columns are advantageous in ensuring high precision data, they are widely available commercially and are easy to use and less time-consuming.
  • Two main approaches one should consider before choosing a RP column are, firstly characterising physiochemical parameters of the packing material such as 1% carbon load, surface area, pore size, particle size and pH.
    • These are some of the important parameters important to control the quality of the column.
  • Second approach is based upon the chromatographic and statistical evaluation of the retention behaviour for a large number of compounds.
  • Kromasil C18 columns are basically RP columns made up of silica-based monomeric type reversed-phase packing material.
TRUSTED_SUSTAINABILITY

Specifications

Column Format HPLC Column
Diameter (Metric) 4 mm
Length (Metric) 125 mm
Particle Size 5 μm
Pore Size 100 Å
USP Type USP L1
Stationary Phase C18 (octadecyl)
Product Line Kromasil
Matrix C18 (octadecyl) Phase
Packing Material Silica-based Monomeric
Column Type Reversed Phase
Phase Reversed Phase
Technique HPLC
Product Type HPLC Column
Show More Show Less

Kromasil is a registered trademark of EKA Chemicals AB

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.