Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Kromasil C8 HPLC Column, 100 Å, 5 μm Particle Size

Catalog No. 111020738 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Length (Metric):
150 mm
250 mm
Diameter (Metric):
4.6 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric)
11-102-0738 250 mm 4.6 mm
11-100-3541 150 mm 4.6 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-102-0738 Supplier MilliporeSigma™ Supelco™ Supplier No. K08670369
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For small molecules and peptides.

Kromasil Classic 100 Å family of products is used to separate and purify molecules of up to about 10,000 Da.
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 4.6 mm
Length (Metric) 250 mm
Particle Size 5 μm
Pore Size 100 Å
Stationary Phase C8 (Octyl)
Product Line Kromasil
Matrix C8 (Octyl) Phase
Phase Reversed Phase
Technique Reverse Phase HPLC

Kromasil is a registered trademark of EKA Chemicals AB

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.