Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Kromasil™ Eternity™ C18 Guard Cartridge

Catalog No. 111008836 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Diameter (Metric) Inner:
3.0 to 4.6 mm
For Use With (Equipment):
Protects 3.0-4.6 mm I.D. Columns
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Inner For Use With (Equipment)
11-100-8836 3.0 to 4.6 mm Protects 3.0-4.6 mm I.D. Columns
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-8836 Supplier MilliporeSigma™ Supelco™ Supplier No. K08971217

Kromasil™ Eternity™ and Eternity™XT can be used in a wider pH window than regular silica-based HPLC materials, giving more flexibility than ever when running analytical applications or optimizing for large-scale applications.

For regular silica-based stationary phases, exposure to extreme pH (especially basic) will have a negative impact on the chemical stability and therefore column lifetime. However, the silica/organosilane surface of the Kromasil™ Eternity™ platform offers a chemical stability that will secure a long-lasting stationary phase, even under tough pH conditions and higher temperatures.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 5/Pk.
Particle Size 5 μm
Length (Metric) 10 m
Pore Size 100 Å
Type Guard Cartridge
Diameter (Metric) Inner 3.0 to 4.6 mm
For Use With (Equipment) Protects 3.0-4.6 mm I.D. Columns
Phase Reversed Phase
For Use With (Application) HPLC
Product Line Kromasil Eternity

Kromasil and Eternity are trademarks of EKA Chemicals AB

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.