Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KRR1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584077
Description
Immunogen sequence: SAIGPFSGLK EVRKVVLDTM KNIHPIYNIK SLMIKRELAK DSELRSQSWE RFLPQFKHKN VNKRKEPKKK TVKKEYTPFP PPQPESQIDK ELA.
Required for 40S ribosome biogenesis. Involved in nucleolar processing of pre-18S ribosomal RNA and ribosome assembly.
Specifications
KRR1 | |
Polyclonal | |
Unconjugated | |
KRR1 | |
2610511F02Rik; AI255219; AI428520; D10Ertd773e; HIV-1 Rev binding protein 2; HIV-1 Rev-binding protein 2; HIV-1 Rev-binding protein 2 homolog; Hrb2; Krr1; KRR1 small subunit processome component homolog; KRR1, small subunit (SSU) processome component, homolog; KRR1, small subunit (SSU) processome component, homolog (yeast); KRR1, small subunit processome component; KRR1, small subunit processome component homolog; KRR-R motif-containing protein 1; Rev interacting protein; rev-interacting protein 1; Rip-1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11103 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q13601 | |
KRR1 | |
Recombinant protein corresponding to Human KRR1. Recombinant protein control fragment (Product #RP-106350). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction