Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KRR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109894
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109894 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109894 Supplier Invitrogen™ Supplier No. PA5109894
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MASPSLERPE KGAGKSEFRN QKPKPENQDE SELLTVPDGW KEPAFSKEDN PRGLLEESSF ATLFPKYREA YLKECWPLVQ KALNEHHVNA TLDLIEGSMT VCTTKKTFDP YIIIRARDLI KLLARSVSFE QAVRILQDDV ACDIIKIGSL VRNKERFVKR RQRLIGPKGS TLKALELLTN CYIMVQGNTV SAIGPFSGLK EVRKVVLDTM KNIHPIYNIK SLMIKRELAK DSELRSQSWE RFLPQFKHKN VNKRKEPKKK TVKKEYTPFP PPQPESQIDK ELASGEYFLK ANQKKRQKME AIKAKQAEAI SKRQEERNKA FIPPKEKPIV KPKEASTETK IDVASIKEKV KKAKNKKLGA LTAEEIALKM EADEKKKKKK K.

Required for 40S ribosome biogenesis. Involved in nucleolar processing of pre-18S ribosomal RNA and ribosome assembly.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KRR1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene KRR1
Gene Accession No. Q13601, Q8BGA5
Gene Alias 2610511F02Rik; AI255219; AI428520; D10Ertd773e; HIV-1 Rev binding protein 2; HIV-1 Rev-binding protein 2; HIV-1 Rev-binding protein 2 homolog; Hrb2; Krr1; KRR1 small subunit processome component homolog; KRR1, small subunit (SSU) processome component, homolog; KRR1, small subunit (SSU) processome component, homolog (yeast); KRR1, small subunit processome component; KRR1, small subunit processome component homolog; KRR-R motif-containing protein 1; Rev interacting protein; rev-interacting protein 1; Rip-1
Gene Symbols KRR1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-381 of human KRR1 (NP_008974.5).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11103, 314830, 52705
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.