Learn More
KRR1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303898A

Description
The recommended shelf life for this product is 1 year from date of receipt.
Required for 40S ribosome biogenesis. Involved in nucleolar processing of pre-18S ribosomal RNA and ribosome assembly.Specifications
KRR1 | |
Polyclonal | |
Unconjugated | |
KRR1 | |
HRB2, RIP-1, HIV-1 Rev binding protein 2, HIV-1 Rev-binding protein 2, KRR-R motif-containing protein 1, KRR1 small subunit processome component homolog, Rev interacting protein, rev-interacting protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11103 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q13601 | |
KRR1 | |
Between 300 and 350 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.