Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRT10 Mouse anti-Human, Unconjugated, Clone: rKRT10/844, Abnova™

Catalog No. 89337933 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-933 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-933 Supplier Abnova Corporation Supplier No. RAB00508
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

KRT10 Monoclonal antibody specifically detects KRT10 in Human samples. It is validated for IHC-P.

Specifications

Antigen KRT10
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone rKRT10/844
Conjugate Unconjugated
Description keratin 10
Gene KRT10
Gene Alias CK10, K10, KPP
Host Species Mouse
Immunogen Original antibody is raised against recombinant protein corresponding to full length human KRT10
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 3858
Target Species Human
Content And Storage Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.