Learn More
KRT14 Mouse anti-Human, Clone: OTI3C7, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF801254

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Cytokeratins (CK) are intermediate filaments of epithelial cells, both in keratinizing tissue (ie. skin) and non-keratinizing cells (ie. mesothelial cells). At least 20 different CKs can be identified. Biochemically, most members of the CK family fall into one of two classes, type I (acidic polypeptides) and type II (basic polypeptides). Belonging to the type I subfamily of low molecular weight keratins and existing in combination with keratin 5, keratin 14 distinguishes stratified epithelial cells from simple epithelial cells and is useful in identification of squamous cell carcinomas. It is considered a prognostic marker in breast carcinomas. At least one member of the acidic family and one member of the basic family is expressed in all epithelial cells.Specifications
KRT14 | |
Monoclonal | |
Unconjugated | |
KRT14 | |
CK14, EBS3, EBS4, K14, NFJ | |
Mouse | |
Affinity Chromatography | |
RUO | |
3861 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI3C7 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P02533 | |
KRT14 | |
Human recombit protein fragment corresponding to amino acids 148-472 of human KRT14 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.