Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRT15 Mouse anti-Human, Unconjugated, Clone: KRT15/2554, Abnova™

Catalog No. 89337773 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-773 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-773 Supplier Abnova Corporation Supplier No. MAB21282
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

KRT15 Monoclonal antibody specifically detects KRT15 in Human samples. It is validated for IHC-Fr, IHC-P.

Specifications

Antigen KRT15
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone KRT15/2554
Conjugate Unconjugated
Description keratin 15
Gene KRT15
Gene Alias CK15, K15, K1CO
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human KRT15
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 3866
Target Species Human
Content And Storage Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.