Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRT15 Rabbit anti-Bovine, Human, Rat, Unconjugated, Clone: KRT15/2103R, Abnova™

Catalog No. 89339523 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-523 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-523 Supplier Abnova Corporation Supplier No. RAB00567
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

KRT15 Monoclonal antibody specifically detects KRT15 in Bovine, Human, Rat samples. It is validated for IHC-P, Flow Cyt.

Specifications

Antigen KRT15
Applications Immunohistochemistry (Paraffin), Flow Cytometry
Classification Monoclonal
Clone KRT15/2103R
Conjugate Unconjugated
Description keratin 15
Gene KRT15
Gene Alias CK15, K15, K1CO
Host Species Rabbit
Immunogen Recombinant protein corresponding to full-length human KRT15
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 3866
Target Species Bovine, Human, Rat
Content And Storage Store at -20 to -80°C
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.