Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRT16 Mouse anti-Human, Unconjugated, Clone: KRT16/1714, Abnova™

Catalog No. 89337938 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-938 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-938 Supplier Abnova Corporation Supplier No. RAB00513
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

KRT16 Monoclonal antibody specifically detects KRT16 in Human samples. It is validated for IHC-P.

Specifications

Antigen KRT16
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone KRT16/1714
Conjugate Unconjugated
Description keratin 16
Gene KRT16
Gene Alias CK16, K16, K1CP, KRT16A, NEPPK
Host Species Mouse
Immunogen Recombinant protein corresponding to C-terminus of human KRT16
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 3868
Target Species Human
Content And Storage Store at 2 to 8°C
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.