Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRT17 Rabbit anti-Human, Unconjugated, Clone: RM351, Abnova™

Catalog No. 89339198 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-198 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-198 Supplier Abnova Corporation Supplier No. MAB21981
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

KRT17 Monoclonal antibody specifically detects KRT17 in Human samples. It is validated for WB-Ce, IHC-P.

Specifications

Antigen KRT17
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone RM351
Conjugate Unconjugated
Description keratin 17
Gene KRT17
Gene Alias K17, PC, PC2, PCHC1
Host Species Rabbit
Immunogen A peptide corresponding to C-terminus of human KRT17
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 3872
Target Species Human
Content And Storage Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.