Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRT18, Mouse anti-Human, Biotin, Clone: C-04, Abnova™

Catalog No. 89999118 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-999-118 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-999-118 Supplier Abnova Corporation Supplier No. MAB16360
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifications

Antigen KRT18
Applications Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone C-04
Conjugate Biotin
Dilution Flow Cytometry (1:500) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide).
Gene keratin 18
Gene Alias CYK18/K18
Gene Symbols KRT18
Host Species Mouse
Immunogen Cytoskeleton preparation of epidermal carcinoma cell line A431.
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3875
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.