Learn More
KRT19 Mouse anti-Human, Clone: OTI8D1, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF804618

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Cytokeratin 19 is part of a subfamily of intermediate filament proteins and are characterized by a remarkable biochemical diversity, represented in human epithelial tissues by at least 20 different polypeptides. Cytokeratins range in molecular weight from 40-68 kDa and isoelectric pH between 4.9 - 7.8. The individual human cytokeratins are numbered 1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial maligcies. The cytokeratin antibodies are not only of assistance in the differential diagnosis of tumors using immunohistochemistry on tissue sections, but are also a useful tool in cytopathology and flow cytometric assays. For example, cytokeratin 19 is not expressed in hepatocytes, therefore, it is useful in the identification of liver metastasis.Specifications
KRT19 | |
Monoclonal | |
Unconjugated | |
KRT19 | |
CK19, K19, K1CS | |
Mouse | |
Affinity Chromatography | |
RUO | |
3880 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI8D1 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P08727 | |
KRT19 | |
Human recombit protein fragment corresponding to amino acids 240-390 of human KRT19 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.