Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRT19 Mouse anti-Human, Unconjugated, Clone: rKRT19/799, Abnova™

Catalog No. 89337949 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-949 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-949 Supplier Abnova Corporation Supplier No. RAB00524
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

KRT19 Monoclonal antibody specifically detects KRT19 in Human samples. It is validated for WB-Ce, IHC-P.

Specifications

Antigen KRT19
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone rKRT19/799
Conjugate Unconjugated
Description keratin 19
Gene KRT19
Gene Alias CK19, K19, K1CS, MGC15366
Host Species Mouse
Immunogen Recombinant protein corresponding to full-length human KRT19
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 3880
Target Species Human
Content And Storage Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.