Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRT20 Mouse anti-Human, Clone: OTI6D5, liquid, TrueMAB™
SDP

Catalog No. 501677376
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-7376 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-7376 Supplier OriGene Supplier No. TA600325
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The protein encoded by this gene is a member of the keratin family. The keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into cytokeratins and hair keratins. The type I cytokeratins consist of acidic proteins which are arranged in pairs of heterotypic keratin chains. This cytokeratin is a major cellular protein of mature enterocytes and goblet cells and is specifically expressed in the gastric and intestinal mucosa. The type I cytokeratin genes are clustered in a region of chromosome 17q12-q21.

Specifications

Antigen KRT20
Classification Monoclonal
Clone OTI6D5
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with <5% trehalose and 0.05% sodium azide
Gene KRT20
Gene Accession No. P35900
Gene Alias CD20, CK-20, CK20, K20, KRT21
Gene Symbols KRT20
Host Species Mouse
Immunogen Full length human recombit protein of human KRT20 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54474
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.