Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRT20 Rabbit anti-Human, Unconjugated, Clone: KRT20/3129R, Abnova™

Catalog No. 89339585 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-585 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-585 Supplier Abnova Corporation Supplier No. RAB00606
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

KRT20 Monoclonal antibody specifically detects KRT20 in Human samples. It is validated for WB-Ce, IHC-P.

Specifications

Antigen KRT20
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone KRT20/3129R
Conjugate Unconjugated
Description keratin 20
Gene KRT20
Gene Alias CD20, CK20, K20, KRT21, MGC35423
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 196-323 of human KRT20
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 54474
Target Species Human
Content And Storage Store at -20 to -80°C
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.