Learn More
Invitrogen™ Krt222 Polyclonal Antibody

Description
Predicted to react with bovine based on sequence homology.
Specifications
Specifications
| Antigen | Krt222 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | Krt222 |
| Gene Accession No. | Q8CCX5, Q8N1A0 |
| Gene Alias | 6330509G02Rik; KA21; keratin 222; keratin 222 pseudogene; keratin 222, type II; keratin-222; Keratin-222 pseudogene; keratin-like protein KRT222; Krt222; KRT222P |
| Gene Symbols | Krt222 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.