Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KRT23 Antibody (OTI3F2), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271780IR
Description
KRT23 Monoclonal specifically detects KRT23 in Human samples. It is validated for Western Blot.Specifications
KRT23 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
KRT23 | |
Recombinant protein fragment corresponding to amino acids 1-270 of human KRT23 (NP_056330) produced in E.coli. | |
0.1 mL | |
25984.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI3F2 | |
Western Blot | |
CK23, CK-23, cytokeratin 23, Cytokeratin-23, DKFZp434G032, HAIK1, hyperacetylation-inducible type I keratin, K23histone deacetylase inducible keratin 23, keratin 23 (histone deacetylase inducible), keratin, type I cytoskeletal 23, keratin-23, MGC26158, type I intermediate filament cytokeratin | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction