Learn More
KRT23 Mouse anti-Human, Clone: OTI3F2, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF808818

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene is a member of the keratin family. The keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into cytokeratins and hair keratins. The type I cytokeratins consist of acidic proteins which are arranged in pairs of heterotypic keratin chains. The type I cytokeratin genes are clustered in a region of chromosome 17q12-q21.Specifications
KRT23 | |
Monoclonal | |
Unconjugated | |
KRT23 | |
CK23, HAIK1, K23 | |
Mouse | |
Affinity Chromatography | |
RUO | |
25984 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI3F2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9C075 | |
KRT23 | |
Human recombit protein fragment corresponding to amino acids 1-270 of human KRT23 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.