Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KRT24 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | KRT24 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15518820
![]() |
Novus Biologicals
NBP15518820UL |
20 μL |
Each for $206.00
|
|
|||||
NBP155188
![]() |
Novus Biologicals
NBP155188 |
100 μL |
Each for $487.50
|
|
|||||
Description
KRT24 Polyclonal specifically detects KRT24 in Human samples. It is validated for Western Blot.Specifications
KRT24 | |
Polyclonal | |
Rabbit | |
Q2M2I5 | |
192666 | |
Synthetic peptides corresponding to KRT24(keratin 24) The peptide sequence was selected from the middle region of KRT24. Peptide sequence GSVNMGSRDLVSGDSRSGSCSGQGRDSSKTRVTKTIVEELVDGKVVSSQV. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
CK-24, Cytokeratin-24, FLJ20261cytokeratin-24, K24, KA24, keratin 24, keratin, type I cytoskeletal 24, keratin-24, MGC138169, MGC138173, Type I keratin-24 | |
KRT24 | |
IgG | |
55 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title