Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KRT24 Monoclonal Antibody (OTI4E8)
GREENER_CHOICE

Catalog No. PIMA526581
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526581 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526581 Supplier Invitrogen™ Supplier No. MA526581
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KRT24 Monoclonal Antibody for Western Blot, IHC (P)

KRT24 encodes a member of the type I (acidic) keratin family, which belongs to the superfamily of intermediate filament (IF) proteins. Keratins are heteropolymeric structural proteins which form the intermediate filament. These filaments, along with actin microfilaments and microtubules, compose the cytoskeleton of epithelial cells. The type I keratin genes are clustered in a region of chromosome 17q12-q21.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KRT24
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4E8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene KRT24
Gene Accession No. Q2M2I5
Gene Alias CK-24; Cytokeratin-24; K24; KA24; keratin 24; keratin 24, type I; keratin, type I cytoskeletal 24; Keratin-24; KRT24; Type I keratin-24
Gene Symbols KRT24
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 139-397 of KRT24 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 192666
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.