Learn More
KRT24 Mouse anti-Human, Clone: OTI2F8, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF803031

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
KRT24 encodes a member of the type I (acidic) keratin family, which belongs to the superfamily of intermediate filament (IF) proteins. Keratins are heteropolymeric structural proteins which form the intermediate filament. These filaments, along with actin microfilaments and microtubules, compose the cytoskeleton of epithelial cells. The type I keratin genes are clustered in a region of chromosome 17q12-q21.Specifications
KRT24 | |
Monoclonal | |
Unconjugated | |
KRT24 | |
K24, KA24 | |
Mouse | |
Affinity Chromatography | |
RUO | |
192666 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI2F8 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q2M2I5 | |
KRT24 | |
Human recombit protein fragment corresponding to amino acids 139-397 of human KRT24 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.