Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KRT24 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554654
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554654 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554654 Supplier Invitrogen™ Supplier No. PA554654
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LDIKTRLEVE IETYRRLLDG EGGGSSFAEF GGRNSGSVNM GSRDLVSGDS RSGSCSGQGR DSSKTRVTKT IVEELVDGKV VSSQVSSIS Highest antigen sequence identity to the following orthologs: Mouse - 62%, Rat - 65%.

KRT24 encodes a member of the type I (acidic) keratin family, which belongs to the superfamily of intermediate filament (IF) proteins. Keratins are heteropolymeric structural proteins which form the intermediate filament. These filaments, along with actin microfilaments and microtubules, compose the cytoskeleton of epithelial cells. The type I keratin genes are clustered in a region of chromosome 17q12-q21.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KRT24
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KRT24
Gene Accession No. Q2M2I5
Gene Alias CK-24; Cytokeratin-24; K24; KA24; keratin 24; keratin 24, type I; keratin, type I cytoskeletal 24; Keratin-24; KRT24; Type I keratin-24
Gene Symbols KRT24
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KRT24. Recombinant protein control fragment (Product #RP-93394).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 192666
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.