Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KRT25 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562888
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562888 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562888 Supplier Invitrogen™ Supplier No. PA562888
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TYCLLIGGDD GACKSGGYKS KDYGSGNVGS QVKDPAKAIV VKKVLEEVDQ RSKILTTRLH SLEEKSQSN Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 87%.

Essential for the proper assembly of type I and type II keratin protein complexes and formation of keratin intermediate filaments in the inner root sheath (irs).
TRUSTED_SUSTAINABILITY

Specifications

Antigen KRT25
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KRT25
Gene Accession No. Q7Z3Z0
Gene Alias ARWH3; CK-25; Cytokeratin-25; K25; K25A; keratin 25; keratin 25, type I; keratin 25A; keratin, type I cytoskeletal 25; Keratin-25; keratin-25A; KRT24IRS1; KRT25; KRT25 (HGNC:30839); KRT25A; type I inner root sheath specific keratin 25 irs1; type I inner root sheath-specific keratin-K25irs1
Gene Symbols KRT25
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KRT25. Recombinant protein control fragment (Product #RP-101612).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 147183
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.