Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KRT25 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584397
Description
Immunogen sequence: RRSCPRPTTG SLRLYGGGTS FGTGNSCGIS GIGSGFS.
Essential for the proper assembly of type I and type II keratin protein complexes and formation of keratin intermediate filaments in the inner root sheath (irs).
Specifications
KRT25 | |
Polyclonal | |
Unconjugated | |
KRT25 | |
ARWH3; CK-25; Cytokeratin-25; K25; K25A; keratin 25; keratin 25, type I; keratin 25A; keratin, type I cytoskeletal 25; Keratin-25; keratin-25A; KRT24IRS1; KRT25; KRT25 (HGNC:30839); KRT25A; type I inner root sheath specific keratin 25 irs1; type I inner root sheath-specific keratin-K25irs1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
147183 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7Z3Z0 | |
KRT25 | |
Recombinant protein corresponding to Human KRT25. Recombinant protein control fragment (Product #RP-106904). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction