Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KRT28 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | KRT28 |
---|---|
Dilution | Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
KRT28 Polyclonal specifically detects KRT28 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
KRT28 | |
Polyclonal | |
Rabbit | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
162605 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:FCSLGTLQFCIDKVNISQNTMSLQFSNGSRHVCLRSGAGSVRPLNGGAGFAGSSACGGSVAGSEFSCALGGGLGSVPGGSHAGGALGNAACIGFAGSEG | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
Unconjugated | |
RUO | |
cytokeratin-28, K25D, K28, keratin 25D, keratin 28, keratin, type I cytoskeletal 28, keratin-28, KRT25D | |
KRT28 | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title