Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRT31 Monoclonal Antibody (OTI2C3), TrueMAB™, OriGene
SDP

Catalog No. 502543958
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3958 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3958 Supplier OriGene Supplier No. CF811097
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KRT31 Monoclonal antibody specifically detects KRT31 in Human samples. It is validated for Western Blot

Specifications

Antigen KRT31
Applications Western Blot
Classification Monoclonal
Clone OTI2C3
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene GLRX3
Gene Accession No. O76003
Gene Alias Ha1; Ha-1; hair keratin; Hair keratin, type I Ha1; hard keratin, type I, 1; hHa1; HKA1; HKA-1; K31; Ka25; keratin 31; keratin 31, type I; keratin complex 1, acidic, gene 1; keratin complex 1, gene 1; keratin, hair, acidic,1; keratin, type I cuticular Ha1; Keratin-31; Kha1; KRT hHa1; Krt1-1; Krt31; KRTHA1; MKHA-1; type I hair keratin KA25
Host Species Mouse
Immunogen Full length human recombinant protein of human GLRX3 produced in HEK293T cell.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10539
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.