Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KRT32 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571400
Description
KRT32 Polyclonal Antibody for Western Blot
The protein encoded by this gene is a member of the keratin gene family. As a type I hair keratin, it is an acidic protein which heterodimerizes with type II keratins to form hair and nails. The type I hair keratins are clustered in a region of chromosome 17q12-q21 and have the same direction of transcription.
Specifications
KRT32 | |
Polyclonal | |
Unconjugated | |
KRT32 | |
HA2; hair keratin; Hair keratin, type I Ha2; hard keratin, type I, 2; HHA2; HKA2; K32; Ka26; keratin 32; keratin 32, type I; keratin, hair, acidic, 2; keratin, type I cuticular Ha2; Keratin-32; KRT32; KRTHA2; type I cuticular hair keratin; type I hair keratin KA26 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
3882 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q14532 | |
KRT32 | |
synthetic peptide directed towards the middle region of human KRT32. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction