Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KRT33A Polyclonal Antibody

Catalog No. PIPA5122048
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5122048 20 μL
PIPA5119245 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5122048 Supplier Invitrogen™ Supplier No. PA5122048
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

KRT33A Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

The protein encoded by this gene is a member of the keratin gene family. It is one of the type I hair keratin genes which are clustered in a region of chromosome 17q12-q21 and have the same direction of transcription. As a type I hair keratin, it is an acidic protein which heterodimerizes with type II keratins to form hair and nails. There are two isoforms of this protein, encoded by two separate genes, KRTHA3A and KRTHA3B. [provided by RefSeq].
TRUSTED_SUSTAINABILITY

Specifications

Antigen KRT33A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene Krt33a
Gene Accession No. O76009, Q8K0Y2
Gene Alias 2310015J09Rik; HA3I; Ha-3I; hair keratin; Hair keratin, type I Ha3-I; hard keratin, type I,3I; HHA3-I; HKA3A; K33A; Ka27; keratin 33A; keratin 33A, type I; keratin, hair, acidic, 3A; keratin, type I cuticular Ha3-I; Keratin-33A; Krt1-3; krt33a; KRTHA3A; ortholog of keratin, hair, acidic, 3A KRTHA3A; RGD1306249; type I hair keratin KA27
Gene Symbols Krt33a
Host Species Rabbit
Immunogen Recombinant protein Keratin 33A. The antigen corresponds to amino acid range 57-363 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3883, 71888
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.