Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cytokeratin 31/33A/33B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100847
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100847 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100847 Supplier Invitrogen™ Supplier No. PA5100847
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total KRT31/33A/33B.

The protein encoded by this gene is a member of the keratin gene family. As a type I hair keratin, it is an acidic protein which heterodimerizes with type II keratins to form hair and nails. The type I hair keratins are clustered in a region of chromosome 17q12-q21 and have the same direction of transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin 31/33A/33B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene KRT33A
Gene Accession No. O76009, Q14525, Q15323, Q61765, Q61897, Q8K0Y2
Gene Alias HA1; Ha-1; HA3I; Ha-3I; HA3II; Ha-3II; hair keratin; hair keratin, type I Ha1; Hair keratin, type I Ha3-I; hair keratin, type I Ha3-II; hard keratin, type I, 1; hard keratin, type I, 3II; hard keratin, type I,3I; hHa1; HHA3-I; HHA3-II; HKA1; HKA3A; HKA3B; K31; K33A; K33B; keratin 31; keratin 31, type I; keratin 33A; keratin 33A, type I; keratin 33B; keratin 33B, type I; keratin, hair, acidic, 3A; keratin, hair, acidic, 3B; keratin, hair, acidic,1; keratin, type I cuticular Ha1; Keratin, type I cuticular Ha3-I; keratin, type I cuticular Ha3-II; Keratin-31; Keratin-33A; Keratin-33B; Krt1-3; KRT31; KRT33A; KRT33B; KRTHA1; KRTHA3A; KRTHA3B; type I hair keratin 3B
Gene Symbols KRT31, KRT33A, KRT33B
Host Species Rabbit
Immunogen A synthesized peptide derived from human KRT33A(Accession O76009), corresponding to amino acid residues E304-N354.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16660, 16671, 303527, 3881, 3883, 3884, 450227, 450228, 71888
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.