Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRT35 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA5139779
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5139779 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5139779 Supplier Thermo Scientific Supplier No. PA5139779
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: MYSSSSCKLP SLSPVARSFS ACSVGLGRSS YRATSCLPAL CLPAGGFATS YSGGGGWFGE GILTGNEKET MQSLNDRLAG YLE

KRT35 is a member of the keratin gene family. This type I hair keratin is an acidic protein which heterodimerizes with type II keratins to form hair and nails. The type I hair keratins are clustered in a region of chromosome 17q12-q21 and have the same direction of transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KRT35
Applications Immunocytochemistry
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KRT35
Gene Accession No. Q92764
Gene Alias AI662400; Ha5; Ha-5; hair keratin; hair keratin acidic 5; hair keratin, type I Ha5; hard keratin, type I, 5; HHA5; HHa5 hair keratin type I intermediate filament; HKA5; K35; Ka30; keratin 35; keratin 35, type I; keratin complex 1, acidic, gene 2; keratin complex 1, acidic, gene 24; keratin complex 1, gene 24; keratin, hair, acidic, 5; keratin, type I cuticular Ha5; Keratin-35; KRT hHa5; Krt1-2; Krt1-24; KRT35; KRTHA5; type I hair keratin KA30; Unknown (protein for MGC:139866); wool (hair) keratin intermediate filament protein
Gene Symbols KRT35
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KRT35
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3886
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.