Learn More
KRT4 Rabbit anti-Human, Unconjugated, Clone: KRT/1877R, Abnova™
Shop All Abnova Corporation ProductsDescription
Specifications
Specifications
| Antigen | KRT4 |
| Applications | Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Flow Cytometry |
| Classification | Monoclonal |
| Clone | KRT/1877R |
| Conjugate | Unconjugated |
| Description | keratin 4 |
| Gene | KRT4 |
| Gene Alias | CK4, CYK4, FLJ31692, K4 |
| Host Species | Rabbit |
| Immunogen | Keratin-enriched preparation from cultured human epithelial cells |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.