Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRT4 Rabbit anti-Human, Unconjugated, Clone: RM355, Abnova™

Catalog No. 89339209 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-209 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-209 Supplier Abnova Corporation Supplier No. MAB21992
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

KRT4 Monoclonal antibody specifically detects KRT4 in Human samples. It is validated for WB-Ce, IHC-P.

Specifications

Antigen KRT4
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone RM355
Conjugate Unconjugated
Description keratin 4
Gene KRT4
Gene Alias CK4, CYK4, FLJ31692, K4
Host Species Rabbit
Immunogen A peptide corresponding to C-terminus of human KRT
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 3851
Target Species Human
Content And Storage Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.