Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRT6A Mouse anti-Human, Unconjugated, Clone: LHK6, Abnova™

Catalog No. 89337762 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-762 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-762 Supplier Abnova Corporation Supplier No. MAB21271
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

KRT6A Monoclonal antibody specifically detects KRT6A in Human samples. It is validated for IHC-P, IF, Flow Cyt.

Specifications

Antigen KRT6A
Applications Immunohistochemistry (Paraffin), Immunofluorescence, Flow Cytometry
Classification Monoclonal
Clone LHK6
Conjugate Unconjugated
Description keratin 6A
Gene KRT6A
Gene Alias CK6A, CK6C, CK6D, K6A, K6C, K6D, KRT6C, KRT6D
Host Species Mouse
Immunogen A synthetic peptide corresponding to C-terminus of human KRT6
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 3853, 3854, 286887
Target Species Human
Content And Storage Store at 4°C
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.